Assay Details
| Gene Symbol: MLL-MLLT10 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF272376.1 | - | 484 | 131 | ||
| AF272384.1 | - | 345 | 133 | |||
| AF272385.1 | - | 166 | 133 | |||
More Information
Set Membership: |
|

MLL-MLLT10 fusion
Genomic Map