Assay Details
Gene Symbol: MLL-GMPS fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF297746.1 | - | 330 | 99 | ||
AF297747.1 | - | 444 | 213 | |||
AF297748.1 | - | 189 | 99 | |||
AF297749.1 | - | 303 | 213 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
