Assay Details
Gene Symbol: CRTC1-MAML2 fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY040324.1 | - | 1 | 193 | 131 | |
AY186997.1 | - | 211 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000321949.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000338797.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000594658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | ENST00000601916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_001098482.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007405.hg.1 | NM_015321.2 | Refseq |
