Assay Details
Gene Symbol: BCL2L14-RUNX1 fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ451147.1 | - | 134 | 88 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BCL2L14-RUNX1 fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ451147.1 | - | 134 | 88 |