Assay Details
| Gene Symbol: SS18-SSX1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | S79325.1 | - | 258 | 83 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007174.hg.1 | ENST00000433276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007174.hg.1 | ENST00000433276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007174.hg.1 | ENST00000433276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007174.hg.1 | ENST00000433276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007174.hg.1 | ENST00000433276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007174.hg.1 | ENST00000433276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007295.hg.1 | ENST00000443473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007295.hg.1 | ENST00000443473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007295.hg.1 | ENST00000443473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007179.hg.1 | ENST00000417503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007179.hg.1 | ENST00000417503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007179.hg.1 | ENST00000417503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000376932.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000376932.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000376932.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000376932.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000376932.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000376932.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000376932.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000412590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000412590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000412590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000412590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000412590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000412590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000509958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000509958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000509958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | ENST00000509958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009726.hg.1 | ENST00000434905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009726.hg.1 | ENST00000434905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009726.hg.1 | ENST00000434905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | ENST00000376909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | ENST00000376909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | ENST00000376909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | ENST00000376909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | ENST00000376909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | ENST00000376909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | ENST00000608568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | ENST00000608568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | ENST00000608568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | ENST00000608568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | ENST00000608568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | ENST00000608568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | NR_073393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | NR_073393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | NR_073393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | NR_073393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | NR_073393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009603.hg.1 | NR_073393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007185.hg.1 | ENST00000441624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007185.hg.1 | ENST00000441624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007185.hg.1 | ENST00000441624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007295.hg.1 | ENST00000443473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007295.hg.1 | ENST00000443473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007295.hg.1 | ENST00000443473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | ENST00000376909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | ENST00000376909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | ENST00000376909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | ENST00000376909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | ENST00000376909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | ENST00000376909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | ENST00000608568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | ENST00000608568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | ENST00000608568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | ENST00000608568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | ENST00000608568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | ENST00000608568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | NR_073393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | NR_073393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | NR_073393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | NR_073393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | NR_073393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009603.hg.1 | NR_073393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000376932.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000376932.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000376932.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000376932.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000376932.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000376932.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000376932.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000412590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000412590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000412590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000412590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000412590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000412590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000509958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000509958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000509958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | ENST00000509958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007169.hg.1 | NR_028366.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007179.hg.1 | ENST00000417503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007179.hg.1 | ENST00000417503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007179.hg.1 | ENST00000417503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007174.hg.1 | ENST00000433276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007174.hg.1 | ENST00000433276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007174.hg.1 | ENST00000433276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007174.hg.1 | ENST00000433276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007174.hg.1 | ENST00000433276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007174.hg.1 | ENST00000433276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007185.hg.1 | ENST00000441624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007185.hg.1 | ENST00000441624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007185.hg.1 | ENST00000441624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009726.hg.1 | ENST00000434905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009726.hg.1 | ENST00000434905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009726.hg.1 | ENST00000434905.1 | Ensembl |

SS18-SSX1 fusion
Genomic Map