Assay Details
Gene Symbol: ARHGEF35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003702.2 | NP_001003702.2 | 2 | 1599 | 100 | |
GenBank mRNA | AK125680.1 | - | 2 | 1544 | 100 | |
BC136868.1 | - | 2 | 1458 | 9020859 | 100 | |
BC136878.1 | - | 2 | 1458 | 9020869 | 100 | |
DQ786206.1 | - | 1 | 217 | 100 | ||
EF560743.1 | - | 2 | 1478 | 100 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07003305.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07030463.hg.1 | TC07003305.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07030463.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07030457.hg.1 | TC07003305.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07030457.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07030457.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000493325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000493325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000493325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | ENST00000493325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | NM_001005328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | NM_001005328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07030458.hg.1 | TC07003305.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07030458.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07030458.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07030459.hg.1 | TC07003305.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07030459.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07030459.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07030462.hg.1 | TC07003305.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07030462.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07030462.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012906.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012906.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012906.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012906.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012906.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012906.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012906.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012906.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012906.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700193127.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700193127.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700193127.hg.1 | TC0700012906.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700012906.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012906.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012906.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012906.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012906.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012906.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012906.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012906.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012906.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700193126.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700193126.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700193126.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700193126.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700193126.hg.1 | TC0700012906.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700193123.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700193123.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700193123.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700193123.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700193123.hg.1 | TC0700012906.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700193121.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700193121.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700193121.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700193121.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700193121.hg.1 | TC0700012906.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700193122.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700193122.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700193122.hg.1 | ENST00000378115.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700193122.hg.1 | NM_001003702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700193122.hg.1 | TC0700012906.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000483587.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000493325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000493325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000493325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | ENST00000493325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | NM_001005328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | NM_001005328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | NR_033942.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012909.hg.1 | NR_033942.1 | Refseq |
