Assay Details
| Gene Symbol: ABCG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022436.2 | NP_071881.1 | 2 - 3 | 408 | 66 | |
| XM_005264480.3 | XP_005264537.1 | 2 - 3 | 429 | 66 | ||
| XM_006712073.3 | XP_006712136.1 | 2 - 3 | 429 | 66 | ||
| XM_006712074.3 | XP_006712137.1 | 2 - 3 | 429 | 66 | ||
| XM_011533024.2 | XP_011531326.1 | 2 - 3 | 429 | 66 | ||
| XM_011533025.2 | XP_011531327.1 | 2 - 3 | 853 | 66 | ||
| XM_011533026.2 | XP_011531328.1 | 2 - 3 | 429 | 66 | ||
| GenBank mRNA | AF312715.2 | - | 2 - 3 | 408 | 66 | |
| AF320293.1 | - | 2 - 3 | 374 | 66 | ||
| AK091997.1 | - | 1 - 2 | 1035 | 66 | ||
| BC111541.1 | - | 1 - 2 | 920 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001794.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC111541.1,XM_011533025.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to nutrient
excretion
response to ionizing radiation
negative regulation of intestinal phytosterol absorption
sterol transport
intestinal cholesterol absorption
cholesterol efflux
response to drug
cholesterol homeostasis
negative regulation of intestinal cholesterol absorption
transmembrane transport
protein binding
ATP binding
ATPase activity
cholesterol transporter activity
ATPase activity, coupled to transmembrane movement of substances
protein heterodimerization activity
response to nutrient
excretion
response to ionizing radiation
negative regulation of intestinal phytosterol absorption
sterol transport
intestinal cholesterol absorption
cholesterol efflux
response to drug
cholesterol homeostasis
negative regulation of intestinal cholesterol absorption
transmembrane transport
protein binding
ATP binding
ATPase activity
cholesterol transporter activity
ATPase activity, coupled to transmembrane movement of substances
protein heterodimerization activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02028817.hg.1 | TC02001794.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02028817.hg.1 | ENST00000260645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02028817.hg.1 | ENST00000405322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02028817.hg.1 | ENST00000543989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02028817.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012417.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200190683.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200190683.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200190683.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200190683.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200190683.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200190683.hg.1 | ENST00000260645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200190683.hg.1 | ENST00000405322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200190683.hg.1 | ENST00000409962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200190683.hg.1 | ENST00000486512.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200190683.hg.1 | NM_022436.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200190683.hg.1 | TC0200012417.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 186136 | - | - |

ABCG5
Genomic Map