Assay Details
Gene Symbol: SSTR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001053.3 | NP_001044.1 | 1 | 1051 | 63 | |
NM_001172560.1 | NP_001166031.1 | 2 | 1034 | 63 | ||
GenBank mRNA | AK308381.1 | - | 1 | 1051 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
