Assay Details
| Gene Symbol: VAPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003574.5 | NP_003565.4 | 4 - 5 | 713 | 114 | |
| XM_011525769.2 | XP_011524071.1 | 4 - 5 | 480 | 114 | ||
| GenBank mRNA | BG488667.1 | - | 4 - 5 | 419 | 114 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000049.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ER to Golgi vesicle-mediated transport
endoplasmic reticulum organization
signal transduction
cell death
sphingolipid biosynthetic process
neuron projection development
protein folding in endoplasmic reticulum
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation by host of viral release from host cell
negative regulation by host of viral genome replication
positive regulation by host of viral genome replication
membrane fusion
protein localization to endoplasmic reticulum
COPII-coated vesicle budding
cell-cell adhesion
signal transducer activity
protein binding
microtubule binding
protein domain specific binding
FFAT motif binding
protein heterodimerization activity
cadherin binding involved in cell-cell adhesion
ER to Golgi vesicle-mediated transport
endoplasmic reticulum organization
signal transduction
cell death
sphingolipid biosynthetic process
neuron projection development
protein folding in endoplasmic reticulum
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation by host of viral release from host cell
negative regulation by host of viral genome replication
positive regulation by host of viral genome replication
membrane fusion
protein localization to endoplasmic reticulum
COPII-coated vesicle budding
cell-cell adhesion
signal transducer activity
protein binding
microtubule binding
protein domain specific binding
FFAT motif binding
protein heterodimerization activity
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000577539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000577539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000577539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000577901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000577901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000577901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000577901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000577901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000577901.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000578314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000578314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000583475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000583475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000583475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000583475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000583475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000583475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000583475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000583879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000583879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000583879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000583879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000583879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000583879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000583879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000583879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000584796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000584796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000584796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000584796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000584796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000584796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000584796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000585042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000585042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000585042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000585042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | ENST00000585042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000544.hg.1 | ENST00000340541.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000544.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000544.hg.1 | TC18000049.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000400000.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000577539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000577539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000577539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000577901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000577901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000577901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000577901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000577901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000577901.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000578314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000578314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000583475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000583475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000583475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000583475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000583475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000583475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000583475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000583879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000583879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000583879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000583879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000583879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000583879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000583879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000583879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000584796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000584796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000584796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000584796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000584796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000584796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000584796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000585042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000585042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000585042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000585042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | ENST00000585042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006656.hg.1 | NM_194434.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145542.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145542.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145542.hg.1 | ENST00000340541.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145542.hg.1 | NM_003574.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145542.hg.1 | TC1800006656.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 139052 | - | - |

VAPA
Genomic Map