Assay Details
| Gene Symbol: MCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197320.1 | NP_001184249.1 | 3 - 4 | 682 | 99 | |
| NM_021960.4 | NP_068779.1 | 2 - 3 | 1141 | 99 | ||
| GenBank mRNA | AF118124.1 | - | 2 - 3 | 982 | 99 | |
| AK294462.1 | - | 3 - 4 | 713 | 99 | ||
| AK297217.1 | - | 3 - 4 | 677 | 99 | ||
| AK300499.1 | - | 3 - 4 | 787 | 99 | ||
| AK304775.1 | - | 3 - 4 | 823 | 99 | ||
| AK312508.1 | - | 2 - 3 | 1013 | 99 | ||
| AK316267.1 | - | 3 - 4 | 969 | 99 | ||
| BC017197.2 | - | 2 - 3 | 987 | 3138465 | 99 | |
| BC071897.1 | - | 2 - 3 | 989 | 6586159 | 99 | |
| BC107735.1 | - | 2 - 3 | 1004 | 4770487 | 99 | |
| BT006640.1 | - | 2 - 3 | 933 | 99 | ||
| FJ917536.1 | - | 3 - 4 | 474 | 99 | ||
| L08246.1 | - | 2 - 3 | 993 | 99 | ||
More Information
Set Membership: |
|

MCL1
Genomic Map