Assay Details
Gene Symbol: SLC25A24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213651.2 | NP_998816.1 | 1 - 2 | 311 | 86 | |
GenBank mRNA | AJ619961.1 | - | 1 - 2 | 243 | 86 | |
AK292567.1 | - | 1 - 2 | 311 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002942.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01045720.hg.1 | TC01002942.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01045720.hg.1 | ENST00000370041.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01045720.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01045719.hg.1 | TC01002942.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01045719.hg.1 | ENST00000264128.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01045719.hg.1 | ENST00000370041.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01045719.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01045719.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212939.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212939.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212939.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212939.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212939.hg.1 | TC0100015160.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100212938.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212938.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212938.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212938.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212938.hg.1 | TC0100015160.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212937.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212937.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212937.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212937.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212937.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212937.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212937.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212937.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212937.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212937.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212937.hg.1 | TC0100015160.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 219882 | - | - |
