Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009634.hg.1 |
ENST00000527787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009634.hg.1 |
ENST00000527787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009649.hg.1 |
ENST00000524692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009649.hg.1 |
ENST00000524692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006595.hg.1 |
ENST00000400154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006595.hg.1 |
ENST00000400154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006831.hg.1 |
ENST00000344638.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006831.hg.1 |
ENST00000344638.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008321.hg.1 |
ENST00000328698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008321.hg.1 |
ENST00000328698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008321.hg.1 |
ENST00000529157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008321.hg.1 |
ENST00000529157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008321.hg.1 |
ENST00000529157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008321.hg.1 |
ENST00000529157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008321.hg.1 |
NM_001002035.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008321.hg.1 |
NM_001002035.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009491.hg.1 |
ENST00000530110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009491.hg.1 |
ENST00000530110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006595.hg.1 |
ENST00000400154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006595.hg.1 |
ENST00000400154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009491.hg.1 |
ENST00000530110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009491.hg.1 |
ENST00000530110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006831.hg.1 |
ENST00000344638.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006831.hg.1 |
ENST00000344638.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009634.hg.1 |
ENST00000527787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009634.hg.1 |
ENST00000527787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009649.hg.1 |
ENST00000524692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009649.hg.1 |
ENST00000524692.1 |
Ensembl |