Assay Details
| Gene Symbol: CTLA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037631.2 | NP_001032720.1 | 2 - 3 | 616 | 74 | |
| GenBank mRNA | U90273.1 | - | 1 - 2 | 350 | 74 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001201.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
immune response
cellular response to DNA damage stimulus
negative regulation of B cell proliferation
T cell costimulation
positive regulation of apoptotic process
negative regulation of regulatory T cell differentiation
B cell receptor signaling pathway
protein binding
adaptive immune response
immune response
cellular response to DNA damage stimulus
negative regulation of B cell proliferation
T cell costimulation
positive regulation of apoptotic process
negative regulation of regulatory T cell differentiation
B cell receptor signaling pathway
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000295854.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000295854.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000295854.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000295854.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000472206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000472206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000472206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000472206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000487393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000487393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | ENST00000487393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018474.hg.1 | TC02001201.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018474.hg.1 | ENST00000295854.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018474.hg.1 | ENST00000302823.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018474.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018474.hg.1 | ENST00000472206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018474.hg.1 | ENST00000487393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018474.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018474.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200174595.hg.1 | ENST00000295854.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174595.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174595.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174595.hg.1 | ENST00000472206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174595.hg.1 | ENST00000487393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174595.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200174595.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200174595.hg.1 | ENST00000295854.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174595.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174595.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174595.hg.1 | ENST00000472206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174595.hg.1 | ENST00000487393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174595.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200174595.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200174595.hg.1 | TC0200010525.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000295854.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000295854.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000295854.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000295854.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000302823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000427473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000472206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000472206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000472206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000472206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000487393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000487393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | ENST00000487393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_001037631.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_005214.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010525.hg.1 | NM_005214.4 | Refseq |

CTLA4
Genomic Map