Assay Details
| Gene Symbol: KCNQ1DN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024627.1 | 1 - 2 | 833 | 92 | ||
| GenBank mRNA | AB039920.1 | - | 1 - 2 | 833 | 92 | |
| AJ404617.2 | - | 1 - 2 | 803 | 92 | ||
| BC098159.1 | - | 1 - 2 | 695 | 92 | ||
| BC098256.1 | - | 1 - 2 | 695 | 92 | ||
| BC098319.1 | - | 1 - 2 | 699 | 92 | ||
| BC098344.1 | - | 1 - 2 | 695 | 92 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | KCNQ1 antisense RNA 1 | KCNQ1-AS1 | -8.6kb | Anti-sense | Chr. 11 - 2840135 - 2861569 on NCBI Build GRCh38 | |
| Downstream | cyclin dependent kinase inhibitor 1C | CDKN1C |
BWCR, BWS, KIP2, WBS, p57, p57Kip2 |
11.3kb | Anti-sense | Chr. 11 - 2883218 - 2885804 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

KCNQ1DN
Genomic Map