Assay Details
Gene Symbol: APCDD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153360.2 | NP_699191.1 | 1 - 2 | 279 | 139 | |
NR_130908.1 | 1 - 2 | 279 | 139 | |||
GenBank mRNA | AK074647.1 | - | 1 - 2 | 234 | 139 | |
BC101758.1 | - | 1 - 2 | 131 | 8069264 | 139 | |
BC101760.1 | - | 1 - 2 | 131 | 8069266 | 139 | |
DA665037.1 | - | 1 - 2 | 279 | 139 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000985.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR20013482.hg.1 | ENST00000371149.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20013482.hg.1 | ENST00000439429.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20013482.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20013482.hg.1 | TC20000985.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20013479.hg.1 | ENST00000371149.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20013479.hg.1 | ENST00000439429.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20013479.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20013479.hg.1 | TC20000985.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20013481.hg.1 | ENST00000371149.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20013481.hg.1 | ENST00000439429.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20013481.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20013481.hg.1 | TC20000985.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000491015.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | ENST00000491015.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009624.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167139.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000167139.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000167139.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167139.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167139.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167139.hg.1 | TC2000009624.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000167139.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000167139.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000167139.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167139.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167139.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167137.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000167137.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000167137.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167137.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167137.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167137.hg.1 | TC2000009624.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000167137.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000167137.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000167137.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167137.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167137.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000491015.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | ENST00000491015.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009624.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167140.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000167140.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000167140.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167140.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167140.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167140.hg.1 | TC2000009624.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000167140.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000167140.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000167140.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167140.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167140.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167136.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167136.hg.1 | TC2000009624.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000167136.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167141.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000167141.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000167141.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167141.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167141.hg.1 | NR_130908.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167141.hg.1 | TC2000009624.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000167141.hg.1 | ENST00000371149.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000167141.hg.1 | ENST00000425773.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000167141.hg.1 | NM_001304787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167141.hg.1 | NM_153360.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000167141.hg.1 | NR_130908.1 | Refseq |
