| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010255.hg.1 |
TC09000788.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010255.hg.1 |
ENST00000291744.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010255.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010257.hg.1 |
TC09000788.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010257.hg.1 |
ENST00000291744.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010257.hg.1 |
ENST00000350339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010257.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010257.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009139.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900162236.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900162236.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900162236.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900162236.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900162236.hg.1 |
TC0900009139.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900162238.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900162238.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900162238.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900162238.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900162238.hg.1 |
ENST00000291744.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900162238.hg.1 |
ENST00000350339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900162238.hg.1 |
NM_004108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900162238.hg.1 |
NM_015837.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900162238.hg.1 |
TC0900009139.hg.1 |
Clariom D TC |