Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057283.hg.1 |
TC01003732.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057283.hg.1 |
ENST00000367188.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057283.hg.1 |
ENST00000543650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057283.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057284.hg.1 |
TC01003732.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057284.hg.1 |
ENST00000367188.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057284.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057285.hg.1 |
TC01003732.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057285.hg.1 |
ENST00000367188.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057285.hg.1 |
ENST00000543650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057285.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017028.hg.1 |
ENST00000367188.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017028.hg.1 |
ENST00000367188.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017028.hg.1 |
ENST00000367188.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017028.hg.1 |
ENST00000367188.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017028.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017028.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017028.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017028.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017028.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229023.hg.1 |
ENST00000367188.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229023.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229023.hg.1 |
ENST00000367188.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229023.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229023.hg.1 |
TC0100017028.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017028.hg.1 |
ENST00000367188.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017028.hg.1 |
ENST00000367188.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017028.hg.1 |
ENST00000367188.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017028.hg.1 |
ENST00000367188.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017028.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017028.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017028.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017028.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017028.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229025.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229025.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229025.hg.1 |
TC0100017028.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100229024.hg.1 |
ENST00000367188.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229024.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229024.hg.1 |
ENST00000367188.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229024.hg.1 |
NM_032833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229024.hg.1 |
TC0100017028.hg.1 |
Clariom D TC |