Assay Details
| Gene Symbol: AP3S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002924.2 | NP_001002924.2 | 1 - 2 | 503 | 96 | |
| GenBank mRNA | BC012614.1 | - | 1 - 2 | 116 | 96 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001002924.2 transcripts.
This assay is in the 5'UTR region of the NM_001002924.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000505423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000505423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000505423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000505423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000507436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000507436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000507436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000509055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000509055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000509055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000514118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000514118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000514118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000515066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000515066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000515066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000515066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000515066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000515066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000515066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | ENST00000515066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009966.hg.1 | ENST00000605743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000316788.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000395548.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000505423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000505423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000505423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000505423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000506430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000507436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000507436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000507436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000509055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000509055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000509055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000514118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000514118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000514118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000515066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000515066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000515066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000515066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000515066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000515066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000515066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | ENST00000515066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008384.hg.1 | NM_001284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009966.hg.1 | ENST00000605743.1 | Ensembl |

AP3S1
Genomic Map