Assay Details
| Gene Symbol: AP3S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002924.2 | NP_001002924.2 | 2 - 3 | 541 | 89 | |
| GenBank mRNA | BC012614.1 | - | 2 - 3 | 154 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001002924.2 transcripts.
Set Membership: |
|

AP3S1
Genomic Map