Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009774.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009774.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009774.hg.1 |
TC20000719.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000160788.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160788.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160788.hg.1 |
TC2000008684.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160788.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160788.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160790.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160790.hg.1 |
TC2000008684.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160790.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
NM_001322.2 |
Refseq |