Assay Details
Gene Symbol: NFE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085386.1 | NP_001078855.1 | 2 - 3 | 677 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3360814 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P25889 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 25906 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 8003 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3360814 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3349912 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3360814 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P25889 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P25889 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P25889 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001085386_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8135227 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8135226 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8135228 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8135225 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8135224 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1560527_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11738921_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11738921_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0700008659.hg.1 | ENST00000420058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008659.hg.1 | ENST00000420058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008659.hg.1 | ENST00000420058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008659.hg.1 | ENST00000420058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008659.hg.1 | NM_001085386.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008659.hg.1 | NM_001085386.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008659.hg.1 | NM_001085386.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008659.hg.1 | NM_001085386.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008659.hg.1 | NM_001085386.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010335.hg.1 | ENST00000558699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010335.hg.1 | ENST00000558699.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700161226.hg.1 | NM_001085386.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700161226.hg.1 | NM_001085386.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700161226.hg.1 | TC0700008659.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1500010335.hg.1 | ENST00000558699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010335.hg.1 | ENST00000558699.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700161225.hg.1 | ENST00000420058.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700161225.hg.1 | NM_001085386.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700161225.hg.1 | ENST00000420058.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700161225.hg.1 | NM_001085386.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700161225.hg.1 | TC0700008659.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700008659.hg.1 | ENST00000420058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008659.hg.1 | ENST00000420058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008659.hg.1 | ENST00000420058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008659.hg.1 | ENST00000420058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008659.hg.1 | NM_001085386.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008659.hg.1 | NM_001085386.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008659.hg.1 | NM_001085386.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008659.hg.1 | NM_001085386.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008659.hg.1 | NM_001085386.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 1560527_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1560527_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560527_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560527_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560527_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560527_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560527_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560527_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560527_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560527_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560527_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560527_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560527_at10 | - | - |
