Assay Details
| Gene Symbol: CEACAM20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102597.2 | NP_001096067.2 | 4 - 5 | 768 | 66 | |
| NM_001102598.2 | NP_001096068.2 | 4 - 5 | 768 | 66 | ||
| NM_001102599.2 | NP_001096069.2 | 4 - 5 | 768 | 66 | ||
| NM_001102600.2 | NP_001096070.2 | 4 - 5 | 768 | 66 | ||
| XM_011526429.2 | XP_011524731.1 | 5 - 6 | 1422 | 66 | ||
| XM_011526430.2 | XP_011524732.1 | 5 - 6 | 1423 | 66 | ||
| XM_011526431.2 | XP_011524733.1 | 5 - 6 | 1424 | 66 | ||
| GenBank mRNA | AY358129.1 | - | 4 - 5 | 1031 | 66 | |
| EU008600.1 | - | 4 - 5 | 768 | 66 | ||
| EU008601.1 | - | 4 - 5 | 768 | 66 | ||
| EU008602.1 | - | 4 - 5 | 768 | 66 | ||
| EU008603.1 | - | 4 - 5 | 768 | 66 | ||
More Information
Set Membership: |
|

CEACAM20
Genomic Map