Assay Details
Gene Symbol: LOC107986752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017012904.1 | XP_016868393.1 | 1 | 1786 | 146 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | family with sequence similarity 20 member C | FAM20C |
DMP-4, DMP4, GEF-CK, RNS |
-177.0 | Sense | Chr. 7 - 192939 - 260774 on NCBI Build GRCh38 |
Downstream | forkhead box L1-like | WI2-2373I1.2 | 26.0kb | Sense | Chr. 7 - 290170 - 294422 on NCBI Build GRCh38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017012904.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
