Assay Details
Gene Symbol: SNORA69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002584.1 | 1 | 70 | 88 | ||
GenBank mRNA | Y11163.1 | - | 1 | 70 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | sosondowah ankyrin repeat domain family member D | SOWAHD | ANKRD58 | -27.2kb | Anti-sense | Chr. X - 119758613 - 119760202 on NCBI Build GRCh38 |
Downstream | UPF3 regulator of nonsense transcripts homolog B (yeast) | UPF3B |
HUPF3B, MRX62, MRXS14, RENT3B, UPF3BP1, UPF3BP2, UPF3BP3, UPF3X, Upf3p-X |
17.8kb | Sense | Chr. X - 119805311 - 119853028 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X001314.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3813 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3814 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916033 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17391619 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916040 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32646836 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916034 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916035 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32646835 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916027 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32646833 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916029 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916037 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916030 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32646849 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32646853 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916042 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32646842 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32646855 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32646849 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916035 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916037 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32646853 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32646835 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916030 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916033 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916034 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916040 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32646842 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916029 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17391619 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916042 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32646836 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32646833 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32646855 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11916027 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11916033 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17391621 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11916032 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11916038 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11916024 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11916027 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11916030 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17391624 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11916026 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11916049 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11916047 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11916051 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11916022 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11916042 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11916045 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11916040 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11916052 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11916029 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NR_002584_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8174716 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8174715 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | ENST00000361575.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | ENST00000361575.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | ENST00000361575.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | ENST00000383895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | ENST00000383895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | ENST00000468844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | ENST00000468844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | ENST00000468844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | ENST00000468844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | ENST00000477403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | ENST00000477403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | ENST00000477403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | ENST00000477403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | NM_001000.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | NM_001000.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | NM_001000.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | NR_002584.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010648.hg.1 | NR_002584.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017655.hg.1 | TC0X001314.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017655.hg.1 | ENST00000383895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017655.hg.1 | NR_002584.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017654.hg.1 | TC0X001314.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017654.hg.1 | ENST00000383895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017654.hg.1 | NR_002584.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172345.hg.1 | ENST00000383895.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172345.hg.1 | ENST00000477403.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172345.hg.1 | NR_002584.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172345.hg.1 | ENST00000383895.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172345.hg.1 | ENST00000477403.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172345.hg.1 | NR_002584.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172345.hg.1 | TC0X00010648.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | ENST00000361575.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | ENST00000361575.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | ENST00000361575.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | ENST00000383895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | ENST00000383895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | ENST00000468844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | ENST00000468844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | ENST00000468844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | ENST00000468844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | ENST00000477403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | ENST00000477403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | ENST00000477403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | ENST00000477403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | NM_001000.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | NM_001000.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | NM_001000.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | NR_002584.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010648.hg.1 | NR_002584.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172346.hg.1 | ENST00000383895.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172346.hg.1 | NR_002584.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172346.hg.1 | ENST00000383895.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172346.hg.1 | NR_002584.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172346.hg.1 | TC0X00010648.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3244883 | Y11163.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3244883 | NR_002584.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3244883 | CR625387.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3244883 | Y11163.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3244883 | NR_002584.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3244883 | CR625387.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3244883 | Y11163.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3244883 | NR_002584.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3244883 | CR625387.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3244883 | Y11163.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3244883 | NR_002584.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3244883 | CR625387.1 | human_transcriptome |
