Assay Details
| Gene Symbol: SNORA52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002585.1 | 1 | 62 | 99 | ||
| GenBank mRNA | AJ609448.1 | - | 1 | 62 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | p53-induced death domain protein 1 | PIDD1 |
LRDD, PIDD |
-2.2kb | Anti-sense | Chr. 11 - 799179 - 809504 on NCBI Build GRCh38 |
| Downstream | patatin like phospholipase domain containing 2 | PNPLA2 |
1110001C14Rik, ATGL, FP17548, PEDF-R, TTS-2.2, TTS2, iPLA2zeta |
7.1kb | Sense | Chr. 11 - 818895 - 825573 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000031.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3816 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27990802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07596093 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07596091 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07596091 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07596093 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27990802 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596130 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596073 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596089 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596063 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596066 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596085 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596141 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596059 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596140 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596129 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16407115 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16407106 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596061 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596074 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16407125 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596093 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596122 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596134 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16407111 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596058 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596075 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16407133 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596143 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596082 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16407120 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596079 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16407131 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596131 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596060 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596076 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596135 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16407098 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596068 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16407137 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16407102 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596091 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16407122 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596087 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16407134 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596127 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07596077 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002585_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7937484 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7937483 | - | - |
| Affymetrix Human Genome U219 Array | 11757140_at | - | - |
| Affymetrix Human Genome U219 Array | 11757140_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000447.hg.1 | ENST00000362915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000447.hg.1 | NR_002585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000447.hg.1 | TC11000031.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000362915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000525722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000525722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000525722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000525722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000526222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000526222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000526222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000526222.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000527517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000527517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000527517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000530398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000530398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000530398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000530398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000530797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000530797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000530797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000530797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000530797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000530797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000530797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000530797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000532004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000532004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000532004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | ENST00000532004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | NM_001004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | NM_001004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | NM_001004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | NM_001004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | NM_001004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | NM_001004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | NM_001004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | NM_001004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006491.hg.1 | NR_002585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000321153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000362915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000525722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000525722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000525722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000525722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000526222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000526222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000526222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000526222.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000527517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000527517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000527517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000530398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000530398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000530398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000530398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000530797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000530797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000530797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000530797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000530797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000530797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000530797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000530797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000532004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000532004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000532004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | ENST00000532004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | NM_001004.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | NM_001004.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | NM_001004.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | NM_001004.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | NM_001004.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | NM_001004.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | NM_001004.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | NM_001004.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006491.hg.1 | NR_002585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100144898.hg.1 | ENST00000362915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100144898.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100144898.hg.1 | NR_002585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100144898.hg.1 | ENST00000362915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100144898.hg.1 | ENST00000524867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100144898.hg.1 | NR_002585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100144898.hg.1 | TC1100006491.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3247679 | NR_002585.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3247679 | AJ609448.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3247679 | NR_002585.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3247679 | AJ609448.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3247679 | NR_002585.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3247679 | AJ609448.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3247679 | NR_002585.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3247679 | AJ609448.1 | human_transcriptome |

SNORA52
Genomic Map