Assay Details
Gene Symbol: RPL31P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002595.1 | 1 | 1350 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Fc fragment of IgG receptor IIb | FCGR2B |
CD32, CD32B, FCG2, FCGR2, IGFR2 |
-5.0kb | Anti-sense | Chr. 1 - 161647243 - 161678654 on NCBI Build GRCh38 |
Downstream | Fc receptor like A | FCRLA |
FCRL, FCRL1, FCRLM1, FCRLX, FCRLb, FCRLc1, FCRLc2, FCRLd, FCRLe, FCRX, FREB |
21.7kb | Anti-sense | Chr. 1 - 161706972 - 161714352 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003450.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00550379 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00550370 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20609849 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20609856 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20609846 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20609847 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20609848 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20609843 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00550367 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00550377 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20609852 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15137557 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20609844 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00550368 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20609850 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 750545 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 34357 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3278137 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3827 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3828 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 60666 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P15317275 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3278137 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3278137 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609838 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609831 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609832 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609844 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609848 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609852 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609856 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00550358 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00550368 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00550377 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00550379 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609846 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609836 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00550366 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00550361 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15137557 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00550370 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609834 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15137557 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609846 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609834 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00550366 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609832 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00550377 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00550368 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609848 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00550379 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00550370 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609831 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609844 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00550358 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609838 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609836 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609852 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00550361 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20609856 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15317275 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00550353 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00550373 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00550351 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00550360 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00550362 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00550371 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00550375 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00550356 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00550358 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00550366 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00550381 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00550377 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00550379 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15137556 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00550348 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00550363 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15317275 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400007415.hg.1 | ENST00000497246.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016192.hg.1 | ENST00000426558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016192.hg.1 | ENST00000426558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016192.hg.1 | ENST00000426558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016192.hg.1 | ENST00000454082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016192.hg.1 | NR_002595.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016192.hg.1 | NR_002595.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016192.hg.1 | NR_002595.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007587.hg.1 | ENST00000431058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01053789.hg.1 | TC01003450.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01053789.hg.1 | ENST00000426558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01053789.hg.1 | NR_002595.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008803.hg.1 | ENST00000458430.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008803.hg.1 | ENST00000458430.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223295.hg.1 | ENST00000426558.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223295.hg.1 | NR_002595.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223295.hg.1 | ENST00000426558.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223295.hg.1 | NR_002595.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223295.hg.1 | TC0100016192.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100223297.hg.1 | ENST00000426558.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223297.hg.1 | NR_002595.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223297.hg.1 | ENST00000426558.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223297.hg.1 | NR_002595.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223297.hg.1 | TC0100016192.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700007587.hg.1 | ENST00000431058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016192.hg.1 | ENST00000426558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016192.hg.1 | ENST00000426558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016192.hg.1 | ENST00000426558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016192.hg.1 | ENST00000454082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016192.hg.1 | NR_002595.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016192.hg.1 | NR_002595.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016192.hg.1 | NR_002595.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008803.hg.1 | ENST00000458430.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008803.hg.1 | ENST00000458430.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223296.hg.1 | ENST00000426558.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223296.hg.1 | ENST00000454082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223296.hg.1 | NR_002595.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223296.hg.1 | ENST00000426558.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223296.hg.1 | ENST00000454082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223296.hg.1 | NR_002595.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223296.hg.1 | TC0100016192.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1400007415.hg.1 | ENST00000497246.1 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054517 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054516 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054516 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054517 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1745171 | NR_002595.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1745171 | BC052594.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1745171 | NR_002595.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1745171 | BC052594.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1745171 | NR_002595.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1745171 | BC052594.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1745171 | NR_002595.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1745171 | BC052594.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1745171 | NR_002595.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1745171 | BC052594.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1745171 | NR_002595.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1745171 | BC052594.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1745171 | NR_002595.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1745171 | BC052594.1 | human_transcriptome |
