| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000382084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000382084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000382084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000382084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000382084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000382084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000382084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000382084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000382086.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000382086.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000382086.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000382086.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000382086.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000382086.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000461833.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000461833.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000461833.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000577714.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000577714.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000577714.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000577714.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000577714.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000577714.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000577714.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
ENST00000621921.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_104232.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_104232.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_104232.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_104232.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_104232.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_104232.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_104232.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008513.hg.1 |
NR_104232.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007133.hg.1 |
ENST00000617561.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012016.hg.1 |
ENST00000382086.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012016.hg.1 |
ENST00000400234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012016.hg.1 |
ENST00000461833.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012016.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012016.hg.1 |
TC22000681.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008504.hg.1 |
ENST00000618747.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000382084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000382084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000382084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000382084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000382084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000382084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000382084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000382084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000382086.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000382086.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000382086.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000382086.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000382086.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000382086.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000461833.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000461833.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000461833.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000577714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000577714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000577714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000577714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000577714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000577714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000577714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
ENST00000621921.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_001450.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_104232.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_104232.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_104232.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_104232.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_104232.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_104232.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_104232.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008513.hg.1 |
NR_104232.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008504.hg.1 |
ENST00000618747.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007133.hg.1 |
ENST00000617561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161281.hg.1 |
TC2200008513.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200161281.hg.1 |
ENST00000382086.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161281.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161281.hg.1 |
ENST00000461833.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161281.hg.1 |
ENST00000577714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161281.hg.1 |
NR_002596.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200161281.hg.1 |
ENST00000382086.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161281.hg.1 |
ENST00000400234.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161281.hg.1 |
ENST00000461833.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161281.hg.1 |
ENST00000577714.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161281.hg.1 |
NR_002596.2 |
Refseq |