Assay Details
| Gene Symbol: NUDT9P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002779.1 | 1 | 947 | 151 | ||
| GenBank mRNA | BC029544.1 | - | 1 | 947 | 151 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 502 | LINC00502 | -89.8kb | Anti-sense | Chr. 10 - 91045808 - 91062159 on NCBI Build GRCh38 | |
| Downstream | polycomb group ring finger 5 | PCGF5 | RNF159 | 9.3kb | Anti-sense | Chr. 10 - 91162403 - 91284331 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NUDT9P1
Genomic Map