Assay Details
| Gene Symbol: GNAS-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002785.2 | 1 - 2 | 371 | 87 | ||
| GenBank mRNA | AJ251759.1 | - | 1 - 2 | 371 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 298 | MIR298 |
MIRN298, hsa-mir-298 |
-30.6kb | Sense | Chr. 20 - 58818226 - 58818313 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927932 | LOC101927932 | 12.9kb | Sense | Chr. 20 - 58863756 - 58888810 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000988.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20013494.hg.1 | ENST00000424094.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013494.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013494.hg.1 | TC20000988.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013493.hg.1 | ENST00000424094.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013493.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013493.hg.1 | TC20000988.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007918.hg.1 | ENST00000616546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007919.hg.1 | ENST00000614281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000385215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000596276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000596276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000596276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000598163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000598163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000598163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000598163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000601795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000601795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000631314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000631314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | ENST00000631314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009638.hg.1 | NR_029844.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000167252.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167252.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167252.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000167252.hg.1 | TC2000009638.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000167252.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167252.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167252.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000167251.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167251.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167251.hg.1 | ENST00000598163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167251.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000167251.hg.1 | TC2000009638.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000167251.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167251.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167251.hg.1 | ENST00000598163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167251.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007918.hg.1 | ENST00000616546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007919.hg.1 | ENST00000614281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000385215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000443966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000596276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000596276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000596276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000598163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000598163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000598163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000598163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000601795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000601795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000631314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000631314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | ENST00000631314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009638.hg.1 | NR_029844.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000167249.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167249.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167249.hg.1 | ENST00000598163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167249.hg.1 | NR_002785.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000167249.hg.1 | TC2000009638.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000167249.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167249.hg.1 | ENST00000424094.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167249.hg.1 | ENST00000598163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167249.hg.1 | NR_002785.2 | Refseq |

GNAS-AS1
Genomic Map