Assay Details
| Gene Symbol: SNORA28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002964.1 | 1 | 64 | 76 | ||
| GenBank mRNA | AJ609462.1 | - | 1 | 64 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105378183 | LOC105378183 | -19.3kb | Sense | Chr. 14 - 103317284 - 103318539 on NCBI Build GRCh38 | |
| Downstream | microtubule affinity regulating kinase 3 | MARK3 |
CTAK1, KP78, PAR1A, Par-1a |
47.4kb | Sense | Chr. 14 - 103385364 - 103503831 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000798.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3709317 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3965 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3964 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 17672 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3709317 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3709317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09526727 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29990021 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09526722 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29990023 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29990016 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09526736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29990015 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29990010 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29990008 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09526732 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09526725 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09526738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29990025 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09526727 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29990025 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29990021 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09526738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09526725 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29990016 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09526736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29990023 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29990015 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29990010 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29990008 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09526722 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09526732 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526727 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526717 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16737346 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526735 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526693 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16737353 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526700 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526698 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526731 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526708 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526736 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526722 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526739 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526719 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526695 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526725 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526710 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526729 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526713 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526721 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526720 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526716 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526702 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09526705 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002964_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7977076 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7977075 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 241843_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11763890_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11763890_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008396.hg.1 | ENST00000606769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008396.hg.1 | NR_002964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008615.hg.1 | ENST00000384585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008615.hg.1 | NR_002964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008615.hg.1 | TC14000798.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400159934.hg.1 | ENST00000606769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400159934.hg.1 | NR_002964.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400159934.hg.1 | ENST00000606769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400159934.hg.1 | NR_002964.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400159934.hg.1 | TC1400008396.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400008396.hg.1 | ENST00000606769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008396.hg.1 | NR_002964.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 241843_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 241843_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241843_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241843_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241843_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241843_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241843_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241843_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241843_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241843_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241843_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241843_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241843_at9 | - | - |
| Human Genome U95 Set | 51379_at | - | - |
| Human Genome U95 Set | 51379_PM_at | - | - |
| Human Genome U133 Set | 241843_at | - | - |
| Human Genome U133 Set | 241843_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3235096 | NR_002964.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3235096 | AJ609462.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3235096 | NR_002964.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3235096 | AJ609462.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3235096 | NR_002964.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3235096 | AJ609462.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3235096 | NR_002964.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3235096 | AJ609462.1 | human_transcriptome |

SNORA28
Genomic Map