Assay Details
| Gene Symbol: SNORA35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002993.1 | 1 | 65 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928437 | LOC101928437 | -1.1mb | Sense | Chr. X - 113042727 - 113520614 on NCBI Build GRCh38 | |
| Downstream | microRNA 764 | MIR764 | hsa-mir-764 | 8.5kb | Sense | Chr. X - 114639435 - 114639519 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000556.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4006 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899047 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899021 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629091 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629071 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899023 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629098 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629062 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629080 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629070 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17388736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629059 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629048 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629089 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629061 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899083 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629083 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899065 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899037 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899051 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629051 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629101 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629052 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629079 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899031 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629074 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899025 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629099 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899073 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899064 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629095 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629086 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629082 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629078 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629093 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629067 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629077 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629066 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629075 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629084 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629064 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899045 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629092 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629102 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629050 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629099 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629101 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629080 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629052 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899023 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629050 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629095 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629079 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629102 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899064 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899047 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629071 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629086 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629062 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629082 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629064 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629048 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629089 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899083 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629075 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629084 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629092 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899073 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629059 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899025 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899021 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899045 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629051 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899037 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629067 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629074 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629078 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629093 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629077 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629083 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899051 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629091 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629070 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629066 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899031 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899065 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629061 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629098 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17388736 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899063 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899036 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899080 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899031 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899075 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899028 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899073 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899069 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899045 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899027 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899044 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899019 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899064 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899049 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899055 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899039 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899021 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899077 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899038 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899022 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899051 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899023 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899030 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899082 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899034 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899054 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899060 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899043 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899025 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17388735 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899053 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899058 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17388738 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899032 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11899067 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002993_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8169460 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8169459 | - | - |
| Affymetrix Human Genome U219 Array | 11757176_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11757175_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11757176_PM_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11757175_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008180.hg.1 | ENST00000386104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008180.hg.1 | NR_002993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007427.hg.1 | TC0X000556.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007427.hg.1 | ENST00000386104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007427.hg.1 | NR_002993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008180.hg.1 | ENST00000386104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008180.hg.1 | NR_002993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155832.hg.1 | ENST00000386104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155832.hg.1 | NR_002993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155832.hg.1 | ENST00000386104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155832.hg.1 | NR_002993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155832.hg.1 | TC0X00008180.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3249351 | NR_002993.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3249351 | NR_002993.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3249351 | NR_002993.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3249351 | NR_002993.1 | human_transcriptome |

SNORA35
Genomic Map