Assay Details
Gene Symbol: FUNDC2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003663.1 | 1 | 1348 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928031 | LOC101928031 | -767.2kb | Sense | Chr. 2 - 83522814 - 83523482 on NCBI Build GRCh38 | |
Downstream | succinate-CoA ligase alpha subunit | SUCLG1 |
GALPHA, MTDPS9, SUCLA1 |
131.3kb | Anti-sense | Chr. 2 - 84423523 - 84459462 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000497.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01325544 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21407890 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15262732 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01325545 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21407888 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21407896 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01325539 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P18250 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 31731 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 45484 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P18250 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_32_P18250 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325544 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407888 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325518 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407859 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407862 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325525 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325501 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325538 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407901 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325504 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407848 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407874 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407851 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325513 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325528 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325520 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325542 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325502 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15262732 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407876 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325541 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407857 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407879 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325529 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325550 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325506 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325526 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407896 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407890 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325500 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407878 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407850 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325534 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325548 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325522 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325543 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407866 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407871 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325510 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325500 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325501 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325528 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325534 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407901 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407850 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407862 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325529 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407874 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325542 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407851 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407879 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407888 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325550 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325504 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325520 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325543 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407871 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325526 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325538 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407866 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325548 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325510 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325544 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407857 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407878 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407848 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15262732 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325522 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407876 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407890 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325506 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407859 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325541 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21407896 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325513 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325518 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325525 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01325502 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P18251 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P18250 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P18250 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325558 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325541 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325518 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325564 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325540 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325500 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325574 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325507 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325501 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325495 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325568 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325502 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325581 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325578 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325546 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15262740 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325582 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325571 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325595 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325508 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325506 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325499 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325591 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325511 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325520 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325526 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325516 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325593 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325566 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325524 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325547 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325550 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325513 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325539 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325515 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15262746 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325522 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15262747 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325576 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325498 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325572 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15262728 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325579 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325561 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325585 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325537 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325554 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325533 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325551 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325599 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325504 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325597 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15262729 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325544 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325535 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325493 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01325538 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NR_003663_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8043042 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8043041 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8043040 | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR02007570.hg.1 | TC02000497.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02007570.hg.1 | ENST00000331369.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02007570.hg.1 | NR_003663.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200156839.hg.1 | ENST00000331369.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200156839.hg.1 | NR_003663.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200156839.hg.1 | ENST00000331369.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200156839.hg.1 | NR_003663.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200156839.hg.1 | TC0200008216.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015891 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015891 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 152722 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1785922 | NR_003663.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1785922 | BC067852.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1785922 | NR_003663.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1785922 | BC067852.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1785922 | NR_003663.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1785922 | BC067852.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1785922 | NR_003663.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1785922 | BC067852.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1785922 | NR_003663.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1785922 | BC067852.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1785922 | NR_003663.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1785922 | BC067852.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1785922 | NR_003663.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1785922 | BC067852.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1785922 | NR_003663.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1785922 | BC067852.1 | human_transcriptome |
