Assay Details
| Gene Symbol: PMS2CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002217.1 | 1 - 2 | 79 | 123 | ||
| GenBank mRNA | AB116525.1 | - | 1 - 2 | 79 | 123 | |
| AK296451.1 | - | 4 - 5 | 754 | 123 | ||
| BC041364.1 | - | 8 - 9 | 834 | 123 | ||
| CR749616.1 | - | 5 - 6 | 418 | 123 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 12 | ZNF12 |
GIOT-3, HZF11, KOX3, ZNF325 |
-28.3kb | Anti-sense | Chr. 7 - 6688433 - 6706935 on NCBI Build GRCh38 |
| Downstream | radial spoke head 10 homolog B2 | RSPH10B2 | 16.2kb | Sense | Chr. 7 - 6754109 - 6798775 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000072.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB116525.1,AK296451.1,CR749616.1 transcripts.
Set Membership: |
|

PMS2CL
Genomic Map