Assay Details
| Gene Symbol: UQCRBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002308.1 | 1 | 654 | 134 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1420 | LINC01420 | Anti-sense | Chr. X - 56729285 - 56817571 on NCBI Build GRCh38 | ||
| Downstream | spindlin family member 3 | SPIN3 |
SPIN-3, TDRD27, bA445O16.1 |
238.8kb | Sense | Chr. X - 56976370 - 56995801 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3345974 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3743 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3742 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 31070 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3345974 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3345974 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32474064 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11760213 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11760208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11760214 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32474057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11760211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32474061 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32474060 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32474058 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32474051 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32474059 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11760216 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32474062 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11760214 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32474051 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11760211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32474062 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11760216 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32474059 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32474058 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11760208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32474057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32474064 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32474061 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11760213 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32474060 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760222 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760178 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760172 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760193 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760210 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760190 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760214 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760189 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760182 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760184 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760180 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760224 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760176 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17351197 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760168 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760220 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760194 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760175 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760192 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760217 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760171 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11760186 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002308_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8173180 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8173179 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039962 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039963 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039962 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039963 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 107653 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1711729 | NR_002308.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1711729 | NR_002308.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1711729 | NR_002308.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1711729 | NR_002308.1 | human_transcriptome |

UQCRBP1
Genomic Map