Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NR_002922_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8107327 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8107326 |
- |
- |
Affymetrix Human Genome U219 Array |
11757145_PM_x_at |
- |
- |
Affymetrix Human Genome U219 Array |
11757144_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11757145_x_at |
- |
- |
Affymetrix Human Genome U219 Array |
11757144_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007335.hg.1 |
TC05000526.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007335.hg.1 |
ENST00000458790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007335.hg.1 |
NR_002922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008319.hg.1 |
ENST00000458790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008319.hg.1 |
NR_002922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008319.hg.1 |
ENST00000458790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008319.hg.1 |
NR_002922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500156946.hg.1 |
ENST00000458790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500156946.hg.1 |
NR_002922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500156946.hg.1 |
ENST00000458790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500156946.hg.1 |
NR_002922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500156946.hg.1 |
TC0500008319.hg.1 |
Clariom D TC |
Human Genome U95 Set |
83234_PM_r_at |
- |
- |
Human Genome U95 Set |
83234_r_at |
- |
- |