Assay Details
| Gene Symbol: SNORA79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003021.2 | 1 | 77 | 112 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | thyroid stimulating hormone receptor | TSHR |
CHNG1, LGR3, hTSHR-I |
-56.4kb | Anti-sense | Chr. 14 - 80954989 - 81146302 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101928504 | LOC101928504 | 18.0kb | Anti-sense | Chr. 14 - 81221218 - 81223380 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001369.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4051 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4050 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424584 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29884996 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424597 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29884989 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885009 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885018 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29884998 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16721196 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885027 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424576 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424582 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424599 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885021 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885000 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424585 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885026 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424575 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885028 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29884987 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885006 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885025 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424590 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29884990 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885002 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16721189 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885003 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424579 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29884999 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29884995 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29884989 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885006 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885003 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424590 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885026 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424599 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424585 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29884987 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29884995 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885009 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29884999 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885002 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885021 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29884996 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424576 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424575 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16721189 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885025 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424584 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885018 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885028 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424579 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424597 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885000 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16721196 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29885027 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09424582 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29884998 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29884990 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16721195 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09424592 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16721196 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09424574 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09424589 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09424564 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09424578 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09424584 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09424561 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16721194 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09424581 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09424567 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09424571 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09424599 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09424594 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16721192 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09424596 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09424569 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09424576 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09424587 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16721190 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400009835.hg.1 | ENST00000408376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009835.hg.1 | NR_003021.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015514.hg.1 | ENST00000408376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015514.hg.1 | NR_003021.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015514.hg.1 | TC14001369.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400172599.hg.1 | ENST00000408376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400172599.hg.1 | NR_003021.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400172599.hg.1 | ENST00000408376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400172599.hg.1 | NR_003021.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400172599.hg.1 | TC1400009835.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400009835.hg.1 | ENST00000408376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009835.hg.1 | NR_003021.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3246206 | NR_003021.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3246206 | NR_003021.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3246206 | NR_003021.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3246206 | NR_003021.2 | human_transcriptome |

SNORA79
Genomic Map