Assay Details
Gene Symbol: EP400NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003290.2 | 8 - 9 | 853 | 107 | ||
GenBank mRNA | AK091234.1 | - | 9 - 10 | 2045 | 107 | |
AK128208.1 | - | 4 - 5 | 1434 | 107 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | E1A binding protein p400 | EP400 |
CAGH32, P400, TNRC12 |
-41.5kb | Sense | Chr. 12 - 131949920 - 132080466 on NCBI Build GRCh38 |
Downstream | DEAD-box helicase 51 | DDX51 | 11.2kb | Anti-sense | Chr. 12 - 132136594 - 132144335 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001053.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
