Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022174.hg.1 |
TC07001466.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022174.hg.1 |
ENST00000457166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022174.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022172.hg.1 |
TC07001466.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022172.hg.1 |
ENST00000457166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022172.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022173.hg.1 |
TC07001466.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022173.hg.1 |
ENST00000457166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022173.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700180939.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700180939.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700180939.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700180939.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700180939.hg.1 |
TC0700011374.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700180941.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700180941.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700180941.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700180941.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700180941.hg.1 |
TC0700011374.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700180942.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700180942.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700180942.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700180942.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700180942.hg.1 |
TC0700011374.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |