Assay Details
| Gene Symbol: TSG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015362.1 | 2 - 3 | 1772 | 75 | ||
| GenBank mRNA | AY314745.1 | - | 2 - 3 | 1772 | 75 | |
| AY314746.1 | - | 2 - 3 | 1772 | 75 | ||
| AY314747.1 | - | 3 - 4 | 1914 | 75 | ||
| AY314748.1 | - | 2 - 3 | 1772 | 75 | ||
| AY314749.1 | - | 1 - 2 | 1987 | 75 | ||
| AY314750.1 | - | 1 - 2 | 1987 | 75 | ||
| AY314751.1 | - | 1 - 2 | 1987 | 75 | ||
| AY314752.1 | - | 2 - 3 | 589 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | EPH receptor A7 | EPHA7 |
EHK-3, EHK3, EK11, HEK11 |
-300.6kb | Anti-sense | Chr. 6 - 93240020 - 93419687 on NCBI Build GRCh38 |
| Downstream | MANEA antisense RNA 1 (head to head) | MANEA-AS1 | 1.8mb | Anti-sense | Chr. 6 - 95560096 - 95577451 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000813.hg.1
Set Membership: |
|

TSG1
Genomic Map