Assay Details
| Gene Symbol: TSG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015362.1 | 8 - 9 | 4376 | 107 | ||
| GenBank mRNA | AY314745.1 | - | 8 - 9 | 4376 | 107 | |
| AY314746.1 | - | 7 - 8 | 4286 | 107 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | EPH receptor A7 | EPHA7 |
EHK-3, EHK3, EK11, HEK11 |
-346.4kb | Anti-sense | Chr. 6 - 93240020 - 93419687 on NCBI Build GRCh38 |
| Downstream | MANEA antisense RNA 1 (head to head) | MANEA-AS1 | 1.8mb | Anti-sense | Chr. 6 - 95560096 - 95577451 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000813.hg.1
Set Membership: |
|

TSG1
Genomic Map