Assay Details
| Gene Symbol: CHKB-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_021492.2 | 1 - 2 | 297 | 83 | ||
| GenBank mRNA | BE393070.1 | - | 1 - 2 | 196 | 83 | |
| BJ989458.1 | - | 1 - 2 | 178 | 83 | ||
| CR456603.1 | - | 1 - 2 | 297 | 83 | ||
| CR980518.1 | - | 1 - 2 | 201 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CHKB-CPT1B readthrough (NMD candidate) | CHKB-CPT1B | CHKL-CPT1B | -27.0 | Anti-sense | Chr. 22 - 50568861 - 50582999 on NCBI Build GRCh38 |
| Downstream | mitogen-activated protein kinase 8 interacting protein 2 | MAPK8IP2 |
IB-2, IB2, JIP2, PRKM8IPL |
16.8kb | Sense | Chr. 22 - 50600685 - 50613978 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000432.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3369079 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3394489 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4411 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4412 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 20362 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 3169 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3369079 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3394489 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3369079 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3394489 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551046 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551049 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551050 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551043 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551040 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32242453 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17299263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551048 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551045 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551056 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32242440 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551051 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551052 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551055 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551055 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551056 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32242440 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32242453 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551045 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551048 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551052 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551040 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551046 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17299263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551050 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551043 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551051 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11551049 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P21355531 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21355531 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11551043 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11551052 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11551045 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17299263 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17299262 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11551056 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11551054 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11551040 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11551051 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11551048 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11551046 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P21355531 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 238557_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | ENST00000609758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | ENST00000609758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | ENST00000609758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | ENST00000609758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | ENST00000609758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007820.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008173.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008173.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008173.hg.1 | TC22000432.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008174.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008174.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008174.hg.1 | TC22000432.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008175.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008175.hg.1 | TC22000432.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200155562.hg.1 | TC2200007820.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200155562.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155562.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155562.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155562.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155558.hg.1 | TC2200007820.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200155558.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155558.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155558.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155558.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155558.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155558.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155563.hg.1 | TC2200007820.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200155563.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155563.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155561.hg.1 | TC2200007820.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200155561.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155561.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155561.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155561.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155557.hg.1 | TC2200007820.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200155557.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155557.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155557.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155557.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155557.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155557.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155559.hg.1 | TC2200007820.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200155559.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155559.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155559.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155559.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155559.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155559.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | ENST00000380711.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | ENST00000609758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | ENST00000609758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | ENST00000609758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | ENST00000609758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | ENST00000609758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | NR_021492.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | NR_110536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007820.hg.1 | NR_110536.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 238557_at | - | - |
| Human Genome U133 Plus 2.0 Array | 238557_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238557_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238557_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238557_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238557_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238557_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238557_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238557_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238557_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238557_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238557_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238557_at4 | - | - |
| Human Genome U95 Set | 85494_at | - | - |
| Human Genome U95 Set | 85494_PM_at | - | - |
| Human Genome U133 Set | 238557_at | - | - |
| Human Genome U133 Set | 238557_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020625 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020625 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3236215 | NR_021492.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3236215 | BJ989458.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3236215 | CR456603.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3236215 | CR980518.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3236215 | BE393070.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3236215 | NR_021492.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3236215 | BJ989458.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3236215 | CR456603.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3236215 | CR980518.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3236215 | BE393070.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3236215 | NR_021492.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3236215 | BJ989458.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3236215 | CR456603.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3236215 | CR980518.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3236215 | BE393070.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3236215 | NR_021492.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3236215 | BJ989458.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3236215 | CR456603.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3236215 | CR980518.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3236215 | BE393070.1 | human_transcriptome |

CHKB-AS1
Genomic Map