Assay Details
| Gene Symbol: VAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014232.2 | NP_055047.2 | 3 - 4 | 373 | 68 | |
| XM_005256775.4 | XP_005256832.1 | 3 - 4 | 703 | 68 | ||
| GenBank mRNA | AJ225044.1 | - | 2 - 3 | 277 | 68 | |
| AK021522.1 | - | 3 - 4 | 355 | 68 | ||
| AK225777.1 | - | 3 - 4 | 373 | 68 | ||
| AK289555.1 | - | 3 - 4 | 346 | 68 | ||
| BC002737.2 | - | 3 - 4 | 342 | 3628842 | 68 | |
| BC019608.1 | - | 3 - 4 | 355 | 4508990 | 68 | |
| BC033870.1 | - | 3 - 4 | 361 | 5203115 | 68 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17002850.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
exocytosis
post-Golgi vesicle-mediated transport
vesicle fusion
neurotransmitter secretion
response to glucose
glutamate secretion
protein transport
synaptic vesicle exocytosis
vesicle-mediated transport
calcium ion regulated exocytosis
regulation of exocytosis
cellular response to insulin stimulus
Golgi to plasma membrane protein transport
eosinophil degranulation
neutrophil degranulation
natural killer cell degranulation
long-term synaptic potentiation
regulation of vesicle-mediated transport
membrane fusion
mucus secretion
positive regulation of intracellular protein transport
regulation of delayed rectifier potassium channel activity
regulation of histamine secretion by mast cell
SNARE binding
SNAP receptor activity
protein binding
calmodulin binding
phospholipid binding
protein C-terminus binding
myosin binding
syntaxin-1 binding
syntaxin binding
protein complex binding
identical protein binding
protein self-association
ion channel binding
calcium-dependent protein binding
protein complex assembly
exocytosis
post-Golgi vesicle-mediated transport
vesicle fusion
neurotransmitter secretion
response to glucose
glutamate secretion
protein transport
synaptic vesicle exocytosis
vesicle-mediated transport
calcium ion regulated exocytosis
regulation of exocytosis
cellular response to insulin stimulus
Golgi to plasma membrane protein transport
eosinophil degranulation
neutrophil degranulation
natural killer cell degranulation
long-term synaptic potentiation
regulation of vesicle-mediated transport
membrane fusion
mucus secretion
positive regulation of intracellular protein transport
regulation of delayed rectifier potassium channel activity
regulation of histamine secretion by mast cell
SNARE binding
SNAP receptor activity
protein binding
calmodulin binding
phospholipid binding
protein C-terminus binding
myosin binding
syntaxin-1 binding
syntaxin binding
protein complex binding
identical protein binding
protein self-association
ion channel binding
calcium-dependent protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17014876.hg.1 | TC17002850.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014876.hg.1 | ENST00000316509.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014876.hg.1 | ENST00000488857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014876.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014879.hg.1 | TC17002850.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014879.hg.1 | ENST00000316509.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014879.hg.1 | ENST00000488857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014879.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014877.hg.1 | TC17002850.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014877.hg.1 | ENST00000316509.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014877.hg.1 | ENST00000488857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014877.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012354.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014875.hg.1 | TC17002850.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014875.hg.1 | ENST00000316509.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014875.hg.1 | ENST00000488857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014875.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205273.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205273.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205273.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205273.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205273.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205273.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205273.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205273.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205273.hg.1 | TC1700012354.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700205266.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205266.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205266.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205266.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205266.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205266.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205266.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205266.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205266.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205266.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205266.hg.1 | TC1700012354.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700205268.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205268.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205268.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205268.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205268.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205268.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205268.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205268.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205268.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205268.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205268.hg.1 | TC1700012354.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700205270.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205270.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205270.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205270.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205270.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205270.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205270.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205270.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205270.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205270.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205270.hg.1 | TC1700012354.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000316509.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000404970.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000481878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | ENST00000488857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | NM_014232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012354.hg.1 | NM_014232.2 | Refseq |

VAMP2
Genomic Map