Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
ENST00000267953.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
ENST00000267953.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
ENST00000267953.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
ENST00000267953.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
ENST00000267953.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
ENST00000267953.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
ENST00000335661.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
ENST00000335661.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
ENST00000335661.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
ENST00000335661.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
ENST00000335661.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
NM_001114735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
NM_001114735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
NM_001114735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
NM_001114735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
NM_001114735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
NM_001114735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
NM_004049.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
NM_004049.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
NM_004049.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
NM_004049.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010184.hg.1 |
NM_004049.3 |
Refseq |