Assay Details
Gene Symbol: HTR7P1 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002774.3 | 1 | 1911 | 81 | |||
GenBank mRNA | U86813.1 | - | LIT2067 serotonin-7 receptor pseudogene |
1 | 851 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | heme binding protein 1 | HEBP1 |
HBP, HEBP |
-1.2kb | Anti-sense | Chr. 12 - 12974864 - 13000309 on NCBI Build GRCh38 |
Downstream | family with sequence similarity 234 member B | FAM234B | KIAA1467 | 41.6kb | Sense | Chr. 12 - 12927680 - 13083449 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 88550 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 136837 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P174572 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P375819 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P375819 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 17011 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 2468 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 38800 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 43489 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P375819 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P174572 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_32_P174572 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P375819 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631558 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631522 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198147 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631545 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198148 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16535543 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631538 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198134 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631540 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198149 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16535539 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631564 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198138 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631530 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16535529 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16535535 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198144 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198135 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198155 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198133 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198161 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631543 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198136 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631555 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631536 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16535545 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198156 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631563 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198154 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198134 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631538 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198161 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631543 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198144 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631558 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198149 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198138 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16535539 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198135 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198156 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198154 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198133 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631530 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198148 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631540 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198155 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198136 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16535543 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16535535 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08198147 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16535529 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631536 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16535545 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631555 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631564 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631545 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631563 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28631522 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P375819 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P174572 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P375819 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198137 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198144 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198136 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198151 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198133 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198159 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16535535 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198141 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198161 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198147 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16535521 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198135 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16535541 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198153 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198143 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16535520 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16535523 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198164 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198139 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198163 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198155 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08198167 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19461372 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NR_002774_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7954074 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7954075 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7954076 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7954071 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 236115_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 216099_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11757237_s_at | - | - |
Affymetrix Human Genome U219 Array | 11757237_PM_s_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1200006904.hg.1 | ENST00000538670.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006904.hg.1 | ENST00000543321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006904.hg.1 | ENST00000543321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006904.hg.1 | ENST00000543321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006904.hg.1 | ENST00000543321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006904.hg.1 | ENST00000543321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006904.hg.1 | ENST00000543321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006904.hg.1 | ENST00000543321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006904.hg.1 | ENST00000624664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006904.hg.1 | ENST00000624664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006904.hg.1 | ENST00000624664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006904.hg.1 | ENST00000624664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006904.hg.1 | NR_002774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006904.hg.1 | NR_002774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006904.hg.1 | NR_002774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006904.hg.1 | NR_002774.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148274.hg.1 | ENST00000624664.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148274.hg.1 | NR_002774.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148274.hg.1 | ENST00000624664.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148274.hg.1 | NR_002774.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148274.hg.1 | TC1200006904.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200006904.hg.1 | ENST00000538670.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006904.hg.1 | ENST00000543321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006904.hg.1 | ENST00000543321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006904.hg.1 | ENST00000543321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006904.hg.1 | ENST00000543321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006904.hg.1 | ENST00000543321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006904.hg.1 | ENST00000543321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006904.hg.1 | ENST00000543321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006904.hg.1 | ENST00000624664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006904.hg.1 | ENST00000624664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006904.hg.1 | ENST00000624664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006904.hg.1 | ENST00000624664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006904.hg.1 | NR_002774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006904.hg.1 | NR_002774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006904.hg.1 | NR_002774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006904.hg.1 | NR_002774.3 | Refseq |
Human Genome U133 Plus 2.0 Array | 216099_at | - | - |
Human Genome U133 Plus 2.0 Array | 216098_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 216098_PM_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 216099_PM_at | - | - |
Human Genome U133A 2.0 Array | 216099_PM_at | - | - |
Human Genome U133A 2.0 Array | 216098_s_at | - | - |
Human Genome U133A 2.0 Array | 216099_at | - | - |
Human Genome U133A 2.0 Array | 216098_PM_s_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NR_002774_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw597035 | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw577184 | - | - |
Human Genome U95 Set | 32403_PM_at | - | - |
Human Genome U95 Set | 32403_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216098_s_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216099_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216099_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216098_s_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216098_s_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216098_s_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216098_s_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216099_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216099_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216099_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216098_s_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216099_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216098_s_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216099_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216098_s_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216099_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216099_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216098_s_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216098_s_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216099_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216099_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216098_s_at3 | - | - |
Human Genome U95 Set | 57621_PM_f_at | - | - |
Human Genome U95 Set | 57620_PM_i_at | - | - |
Human Genome U95 Set | 57620_i_at | - | - |
Human Genome U95 Set | 57621_f_at | - | - |
Human Genome U133 Set | 216099_PM_at | - | - |
Human Genome U133 Set | 216099_at | - | - |
Human Genome U133 Set | 216098_PM_s_at | - | - |
Human Genome U133 Set | 216098_s_at | - | - |
Human Genome U133 Set | 236115_at | - | - |
Human Genome U133 Set | 236115_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A048399 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A048400 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A048400 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A048399 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2164522 | U86813.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2164522 | NR_002774.3 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2164522 | AK125093.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2164522 | U86813.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2164522 | NR_002774.3 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2164522 | AK125093.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2164522 | U86813.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2164522 | NR_002774.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2164522 | AK125093.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2164522 | U86813.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2164522 | NR_002774.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2164522 | AK125093.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2164522 | U86813.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2164522 | NR_002774.3 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2164522 | AK125093.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2164522 | U86813.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2164522 | NR_002774.3 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2164522 | AK125093.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2164522 | U86813.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2164522 | NR_002774.3 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2164522 | AK125093.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2164522 | U86813.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2164522 | NR_002774.3 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2164522 | AK125093.1 | human_transcriptome |
