Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HOXA11 antisense RNA | HOXA11-AS |
HOXA-AS5, HOXA11-AS1, HOXA11AS, HOXA11S, NCRNA00076 |
Sense | Chr. 7 - 27185408 - 27189293 on NCBI Build GRCh38 | |
| Downstream | homeobox A13 | HOXA13 |
HOX1, HOX1J |
4.1kb | Anti-sense | Chr. 7 - 27196880 - 27200106 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map