Assay Details
| Gene Symbol: MNX1 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY927465.1 | - | LIT3487 AY927465 |
1 - 2 | 84 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MNX1 antisense RNA 2 | MNX1-AS2 | Anti-sense | Chr. 7 - 157005863 - 157007120 on NCBI Build GRCh38 | ||
| Downstream | MNX1 antisense RNA 1 (head to head) | MNX1-AS1 | CCAT5 | 1.8kb | Anti-sense | Chr. 7 - 157010857 - 157016424 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000252971.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000252971.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000252971.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000252971.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000252971.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000252971.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000252971.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000425745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000425745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000425745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000428439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000428439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000428439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000428439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000428439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000469500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000469500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000474448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000474448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000474448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000474448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000474448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000474448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000479817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000479817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000543409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000543409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000543409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000543409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000543409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000543409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000543409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000605400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000605400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000605400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | ENST00000605400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | NM_001165255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | NM_001165255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | NM_001165255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | NM_001165255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | NM_001165255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | NM_001165255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | NM_001165255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | NM_005515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | NM_005515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | NM_005515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | NM_005515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | NM_005515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | NM_005515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013231.hg.1 | NM_005515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000252971.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000252971.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000252971.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000252971.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000252971.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000252971.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000252971.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000425745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000425745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000425745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000428439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000428439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000428439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000428439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000428439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000469500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000469500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000474448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000474448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000474448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000474448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000474448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000474448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000479817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000479817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000543409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000543409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000543409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000543409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000543409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000543409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000543409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000605400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000605400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000605400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | ENST00000605400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | NM_001165255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | NM_001165255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | NM_001165255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | NM_001165255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | NM_001165255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | NM_001165255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | NM_001165255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | NM_005515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | NM_005515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | NM_005515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | NM_005515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | NM_005515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | NM_005515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013231.hg.1 | NM_005515.3 | Refseq |

MNX1
Genomic Map