Assay Details
| Gene Symbol: SLC4A2 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY927498.1 | - | LIT3512 AY927498 |
1 | 149 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cyclin dependent kinase 5 | CDK5 |
LIS7, PSSALRE |
-4.0kb | Anti-sense | Chr. 7 - 151053812 - 151057965 on NCBI Build GRCh38 |
| Downstream | Fas activated serine/threonine kinase | FASTK | FAST | 14.5kb | Anti-sense | Chr. 7 - 151076621 - 151080883 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SLC4A2
Genomic Map