Assay Details
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | defective in cullin neddylation 1 domain containing 2 | DCUN1D2 | C13orf17 | -21.0kb | Sense | Chr. 13 - 113455819 - 113492047 on NCBI Build GRCh38 |
Downstream | transcription factor Dp-1 | TFDP1 |
DILC, DP1, DRTF1, Dp-1 |
70.2kb | Anti-sense | Chr. 13 - 113584688 - 113641473 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
