Assay Details
| Gene Symbol: LSS | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY927524.1 | - | LIT3533 AY927524 |
3 - 4 | 290 | 59 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | spermatogenesis and centriole associated 1-like | SPATC1L | C21orf56 | -31.4kb | Sense | Chr. 21 - 46161148 - 46185159 on NCBI Build GRCh38 |
| Downstream | MCM3AP antisense RNA 1 | MCM3AP-AS1 |
C21orf85, MCM3AP-AS, MCM3APAS, MCM3APASB, NCRNA00031 |
9.7kb | Anti-sense | Chr. 21 - 46229231 - 46251701 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LSS
Genomic Map