Assay Details
| Gene Symbol: PISD | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY927618.1 | - | LIT3606 AY927618 |
2 - 3 | 215 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 7109 | MIR7109 | hsa-mir-7109 | -378.0 | Sense | Chr. 22 - 31621467 - 31621531 on NCBI Build GRCh38 |
| Downstream | proline rich 14 like | PRR14L | C22orf30 | 57.5kb | Sense | Chr. 22 - 31681347 - 31750176 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

PISD
Genomic Map